Assay Details
Gene Symbol: GPAA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027973350.1 | XP_027829151.1 | 8 - 9 | 1273 | 63 | |
XM_027973351.1 | XP_027829152.1 | 8 - 9 | 1174 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() |
