Assay Details
Gene Symbol: LOC101120058 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004005253.4 | XP_004005302.3 | 10 - 11 | 1105 | 88 | |
XM_027964322.1 | XP_027820123.1 | 9 - 10 | 1033 | 88 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
