Assay Details
Gene Symbol: LOC101120058 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004005253.4 | XP_004005302.3 | 1 - 2 | 181 | 94 | |
XM_027964322.1 | XP_027820123.1 | 1 - 2 | 181 | 94 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
