Assay Details
| Gene Symbol: MGAT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004010499.4 | XP_004010548.2 | 1 | 1519 | 85 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

MGAT2| Gene Symbol: MGAT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004010499.4 | XP_004010548.2 | 1 | 1519 | 85 | |
Set Membership: |
|