Assay Details
Gene Symbol: TAF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004014182.4 | XP_004014231.1 | 6 - 7 | 2888 | 60 | |
XM_012188167.3 | XP_012043557.1 | 5 - 6 | 2593 | 60 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
