Assay Details
| Gene Symbol: ENPP4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004018862.4 | XP_004018911.1 | 2 - 3 | 1079 | 84 | |
| XM_027959131.1 | XP_027814932.1 | 2 - 3 | 1203 | 84 | ||
| XM_027959132.1 | XP_027814933.1 | 2 - 3 | 934 | 84 | ||
More Information
Set Membership: |
|

ENPP4
Genomic Map