Assay Details
| Gene Symbol: MINPP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004020010.4 | XP_004020059.3 | 1 - 2 | 645 | 115 | |
| XM_012102486.3 | XP_011957876.3 | 1 - 2 | 645 | 115 | ||
More Information
Set Membership: |
|

MINPP1
Genomic Map