Assay Details
Gene Symbol: STYXL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015104216.2 | XP_014959702.1 | 7 - 8 | 834 | 84 | |
XM_027961796.1 | XP_027817597.1 | 7 - 8 | 958 | 84 | ||
XM_027961797.1 | XP_027817598.1 | 5 - 6 | 624 | 84 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
