Assay Details
| Gene Symbol: LOC101120466 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027964020.1 | XP_027819821.1 | 13 - 14 | 1679 | 72 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101120466| Gene Symbol: LOC101120466 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027964020.1 | XP_027819821.1 | 13 - 14 | 1679 | 72 | |
Set Membership: |
|