Assay Details
| Gene Symbol: MRPL57 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015097989.2 | XP_014953475.2 | 2 | 530 | 64 | |
| Gene Symbol: LOC114117223 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027975612.1 | XP_027831413.1 | 1 | 2042 | 64 | |
More Information
Set Membership: |
|

MRPL57
Genomic Map