Assay Details
| Gene Symbol: RALA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004008002.4 | XP_004008051.1 | 4 - 5 | 771 | 96 | |
| XM_027968754.1 | XP_027824555.1 | 4 - 5 | 665 | 96 | ||
More Information
Set Membership: |
|

RALA
Genomic Map