Assay Details
Gene Symbol: DFFA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004013730.4 | XP_004013779.1 | 4 - 5 | 719 | 63 | |
XM_012187337.3 | XP_012042727.1 | 4 - 5 | 719 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() |
