Assay Details
Gene Symbol: LOC101120029 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004019629.3 | XP_004019678.2 | 3 - 4 | 509 | 80 | |
XM_027959689.1 | XP_027815490.1 | 3 - 4 | 361 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
