Assay Details
Gene Symbol: LOC114110255 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027960217.1 | XP_027816018.1 | 2 - 3 | 982 | 109 |
Gene Symbol: LOC101119597 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027960410.1 | XP_027816211.1 | 2 - 3 | 278 | 109 |

More Information
Set Membership: |
![]() ![]() |
