Assay Details
| Gene Symbol: PACSIN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015102662.2 | XP_014958148.2 | 8 - 9 | 2598 | 71 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

PACSIN1| Gene Symbol: PACSIN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015102662.2 | XP_014958148.2 | 8 - 9 | 2598 | 71 | |
Set Membership: |
|