Assay Details
Gene Symbol: RSPH6A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015100604.2 | XP_014956090.2 | 4 - 5 | 1893 | 59 | |
XM_027978495.1 | XP_027834296.1 | 4 - 5 | 1893 | 59 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
