Assay Details
Gene Symbol: MARCHF9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027967508.1 | XP_027823309.1 | 3 - 4 | 885 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
