Assay Details
Gene Symbol: ANAPC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027966067.1 | XP_027821868.1 | 3 - 4 | 916 | 56 | |
XM_027966068.1 | XP_027821869.1 | 3 - 4 | 886 | 56 |

More Information
Set Membership: |
![]() ![]() ![]() |
