Assay Details
Gene Symbol: CCDC106 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027978762.1 | XP_027834563.1 | 5 - 6 | 1727 | 91 | |
XM_027978763.1 | XP_027834564.1 | 5 - 6 | 1049 | 91 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
