Assay Details
Gene Symbol: P2RY1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004003238.4 | XP_004003287.1 | 1 - 2 | 1225 | 90 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
