Assay Details
| Gene Symbol: SLC22A5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027969943.1 | XP_027825744.1 | 2 - 3 | 537 | 68 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SLC22A5| Gene Symbol: SLC22A5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027969943.1 | XP_027825744.1 | 2 - 3 | 537 | 68 | |
Set Membership: |
|