Assay Details
Gene Symbol: ANAPC15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015101069.2 | XP_014956555.1 | 4 - 5 | 591 | 114 | |
XM_027979480.1 | XP_027835281.1 | 4 - 5 | 449 | 114 | ||
XM_027979482.1 | XP_027835283.1 | 4 - 5 | 418 | 114 | ||
XM_027979483.1 | XP_027835284.1 | 4 - 5 | 412 | 114 | ||
XM_027979484.1 | XP_027835285.1 | 4 - 5 | 586 | 114 | ||
XM_027979485.1 | XP_027835286.1 | 4 - 5 | 394 | 114 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
