Assay Details
| Gene Symbol: TMPRSS11A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012180072.3 | XP_012035462.1 | 7 - 8 | 994 | 75 | |
| XM_012180073.3 | XP_012035463.1 | 6 - 7 | 955 | 75 | ||
| XM_012180074.3 | XP_012035464.1 | 7 - 8 | 952 | 75 | ||
More Information
Set Membership: |
|

TMPRSS11A
Genomic Map