Assay Details
Gene Symbol: CHI3L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004013541.3 | XP_004013590.1 | 3 - 4 | 393 | 63 | |
XM_012186898.3 | XP_012042288.1 | 3 - 4 | 396 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
