Assay Details
| Gene Symbol: RPL32 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004018540.3 | XP_004018589.1 | 2 - 3 | 203 | 63 | |
| GenBank mRNA | AF190579.1 | - | 1 - 2 | 86 | 63 | |
More Information
Set Membership: |
|

RPL32
Genomic Map