Assay Details
Gene Symbol: RAMAC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004017775.3 | XP_004017824.1 | 3 - 4 | 338 | 70 | |
XM_012098521.2 | XP_011953911.1 | 4 - 5 | 361 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
