Assay Details
| Gene Symbol: SGF29 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012104338.3 | XP_011959728.1 | 3 - 4 | 311 | 59 | |
| XM_012104339.3 | XP_011959729.1 | 4 - 5 | 295 | 59 | ||
More Information
Set Membership: |
|

SGF29
Genomic Map