Assay Details
| Gene Symbol: NOL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027978058.1 | XP_027833859.1 | 3 - 4 | 826 | 81 | |
| XM_027978060.1 | XP_027833861.1 | 3 - 4 | 1920 | 81 | ||
| XM_027978061.1 | XP_027833862.1 | 4 - 5 | 874 | 81 | ||
| XM_027978062.1 | XP_027833863.1 | 3 - 4 | 798 | 81 | ||
| XM_027978063.1 | XP_027833864.1 | 3 - 4 | 956 | 81 | ||
More Information
Set Membership: |
|

NOL3
Genomic Map