Assay Details
| Gene Symbol: C12H1orf21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012187716.3 | XP_012043106.1 | 3 - 4 | 469 | 113 | |
| XM_015099359.2 | XP_014954845.1 | 3 - 4 | 617 | 113 | ||
More Information
Set Membership: |
|

C12H1orf21
Genomic Map