Assay Details
Gene Symbol: PAX7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027965643.1 | XP_027821444.1 | 4 - 5 | 689 | 78 | |
XM_027965644.1 | XP_027821445.1 | 4 - 5 | 683 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
