Assay Details
Gene Symbol: LOC101114198 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004011124.4 | XP_004011173.2 | 26 - 27 | 4279 | 119 | |
XM_012182449.3 | XP_012037839.2 | 26 - 27 | 4282 | 119 | ||
XM_012182451.3 | XP_012037841.2 | 26 - 27 | 4283 | 119 |
Gene Symbol: LOC114116228 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027972738.1 | XP_027828539.1 | 4 - 5 | 572 | 119 |

More Information
Set Membership: |
![]() ![]() ![]() |
