Assay Details
| Gene Symbol: VNN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004011339.4 | XP_004011388.3 | 3 - 4 | 549 | 102 | |
| GenBank mRNA | KC425611.1 | - | 3 - 4 | 555 | 102 | |
More Information
Set Membership: |
|

VNN2
Genomic Map