Assay Details
Gene Symbol: MSC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004011735.3 | XP_004011784.1 | 1 - 2 | 824 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
