Assay Details
Gene Symbol: POP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004015128.3 | XP_004015177.1 | 4 - 5 | 611 | 65 | |
XM_012190086.2 | XP_012045476.1 | 4 - 5 | 588 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
