Assay Details
| Gene Symbol: ACAD8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027979828.1 | XP_027835629.1 | 8 - 9 | 2867 | 57 | |
| XM_027979829.1 | XP_027835630.1 | 9 - 10 | 1144 | 57 | ||
More Information
Set Membership: |
|

ACAD8
Genomic Map