Assay Details
Gene Symbol: NDUFB10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004023656.4 | XP_004023705.4 | 2 - 3 | 369 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
