Assay Details
| Gene Symbol: PAOX | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027960288.1 | XP_027816089.1 | 3 - 4 | 949 | 85 | |
| XM_027960290.1 | XP_027816091.1 | 3 - 4 | 949 | 85 | ||
| XM_027960291.1 | XP_027816092.1 | 3 - 4 | 949 | 85 | ||
| XM_027960292.1 | XP_027816093.1 | 3 - 4 | 949 | 85 | ||
More Information
Set Membership: |
|

PAOX
Genomic Map