Assay Details
| Gene Symbol: PRAG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027962715.1 | XP_027818516.1 | 3 - 4 | 2358 | 101 | |
| XM_027962716.1 | XP_027818517.1 | 3 - 4 | 2542 | 101 | ||
| XM_027962717.1 | XP_027818518.1 | 3 - 4 | 2844 | 101 | ||
More Information
Set Membership: |
|

PRAG1
Genomic Map