Assay Details
Gene Symbol: CPZ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027971269.1 | XP_027827070.1 | 7 - 8 | 1277 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
