Assay Details
Gene Symbol: LOC101115808 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027974000.1 | XP_027829801.1 | 25 - 26 | 3960 | 110 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
