Assay Details
Gene Symbol: EMC10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027977528.1 | XP_027833329.1 | 5 - 6 | 672 | 77 | |
XM_027977529.1 | XP_027833330.1 | 5 - 6 | 672 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
