Assay Details
| Gene Symbol: SHROOM2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027963343.1 | XP_027819144.1 | 4 - 5 | 2882 | 95 | |
| XM_027963344.1 | XP_027819145.1 | 4 - 5 | 2882 | 95 | ||
| XM_027963345.1 | XP_027819146.1 | 5 - 6 | 2913 | 95 | ||
More Information
Set Membership: |
|

SHROOM2
Genomic Map