Assay Details
| Gene Symbol: DUSP21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015104757.2 | XP_014960243.1 | 1 | 91 | 77 | |
| XM_027963266.1 | XP_027819067.1 | 1 | 91 | 77 | ||
More Information
Set Membership: |
|

DUSP21
Genomic Map