Assay Details
Gene Symbol: GPBP1L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027969368.1 | XP_027825169.1 | 8 - 9 | 2100 | 102 | |
XM_027969377.1 | XP_027825178.1 | 8 - 9 | 1981 | 102 | ||
XM_027969386.1 | XP_027825187.1 | 8 - 9 | 2013 | 102 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
