Assay Details
Gene Symbol: CCNYL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004004867.4 | XP_004004916.3 | 1 - 2 | 569 | 70 | |
XM_004004868.4 | XP_004004917.2 | 1 - 2 | 571 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() |
