Assay Details
| Gene Symbol: PPP1R36 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004011047.4 | XP_004011096.1 | 4 - 5 | 351 | 69 | |
| XM_012181830.2 | XP_012037220.1 | 4 - 5 | 342 | 69 | ||
| XM_012181833.3 | XP_012037223.1 | 4 - 5 | 351 | 69 | ||
More Information
Set Membership: |
|

PPP1R36
Genomic Map