Assay Details
| Gene Symbol: PRSS42 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027957976.1 | XP_027813777.1 | 2 - 3 | 534 | 102 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

PRSS42| Gene Symbol: PRSS42 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027957976.1 | XP_027813777.1 | 2 - 3 | 534 | 102 | |
Set Membership: |
|