Assay Details
Gene Symbol: UPF3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015098281.2 | XP_014953767.2 | 8 - 9 | 1049 | 88 | |
XM_027973835.1 | XP_027829636.1 | 7 - 8 | 843 | 88 |

More Information
Set Membership: |
![]() ![]() ![]() |
