Assay Details
Gene Symbol: LOC114116722 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027973912.1 | XP_027829713.1 | 5 - 6 | 294 | 88 |
Gene Symbol: LOC101122608 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027973511.1 | XP_027829312.1 | 10 - 11 | 88 | ||
GenBank mRNA | FJ958364.1 | - | 10 - 11 | 639 | 88 |
