Assay Details
Gene Symbol: LOC114116722 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027973912.1 | XP_027829713.1 | 2 - 3 | 58 | 97 |
Gene Symbol: LOC101122608 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027973511.1 | XP_027829312.1 | 7 - 8 | 604 | 97 | |
GenBank mRNA | FJ958364.1 | - | 7 - 8 | 403 | 97 |
